Learn More
Grant Instruments™ Dimpled Mat for Platform Shaker
For use with PMR-30 Compact Fixed Angle Platform Rocker. Grant Instruments™ Dimpled Mat keeps vessels from sliding during operation.
Brand:  Grant Instruments™ PDM
Product Code. 11441038
  • 44.52 € / Each
Estimated Shipment: 02-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • For use with PMR-30 Compact Fixed Angle Platform Rocker.
  • Keeps vessels from sliding during operation.
  • Dimensions: 215 x 215mm
Specifications
Specifications
PMR-30