missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bode Chemie™ Dip Slides Combi
Monitor the hygiene status within the scope of GMP (Good Manufacturing Practices) and HACCP (Hazard Analysis of Critical Control Points) with Bode Chemie™ Dip Slides Combi.
Brand: Bode Chemie™ 900130
This item is not returnable.
View return policy
Description
- 2-chamber system (2 diff. agar media per slide) for separate detection of aerobic bacteria, yeasts and fungi
- Culture media stored at room temperature (approx. 20°C), protected against draft and light
- Easy handling makes test suitable for staff without microbiological training
Specifications
Light yellow TTC & pink Rose Bengal agar. | |
Microbiology Media | |
Incubator or room temperature | |
2 x 10 pcs. in a box |
Filamentous fungi / yeasts and bacteria | |
Sterile | |
Incubation in the incubator (27 °C – 30 °C)for bacteria: 1 – 2 daysfor yeasts and fungi: 3 days | |
2-chamber |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Bode Chemie™ Dip Slides Combi >
Spot an opportunity for improvement?Share a Content Correction