missing translation for 'onlineSavingsMsg'
Learn More

Bode Chemie™ Dismozon™ Plus

Dismozon™ Plus Oxygen-active surface disinfectant cleaner. The granules especially for sensitive surfaces and for final disinfection.

Brand:  Bode Chemie™ 9811872


Product Code. 16504030

  • 295.00 € / Pack of 100
Estimated Shipment: 09-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Comprehensive spectrum of activity including virucidal activity and bacterial spores
  • Low residue
  • Extraordinarily wide material compatibility
  • RKI-listed (area A/B)
  • Practical application in a dosing bag
  • Required spectrum of activity SARS-CoV-2: limited virucidal activity
Specifications

Specifications

Granule
Dosing Bag
Disinfecting Cleaning of Washable Surfaces in Medical Areas and Industry
16 g
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • X100 Dismozon plus (100 x 16gr Sachets)

Signal Word
  • Danger

Hazard Category
  • Organic peroxide Type G
  • Skin corrosion/irritation Category 1 A

Hazard Statement
  • H242-Heating may cause a fire.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402-Store in a dry place.
  • P411-Store at temperatures not exceeding °C/°F.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : magnesium monoperoxyphthalate hexahydrate, tridecanol, amines

SDS
Documents

Documents

Certificates
Promotions

Promotions