Learn More
Brady™ Disposable Chemical Spill Kit
Product Code. 15995171
Click to view available options
:
Color: Green
Product Code. 15995171
Brand: Brady™ 813868
Please to purchase this item. Need a web account? Register with us today!
Portable spill kit that comes in a green, disposable dispenser box.
  • Portable
  • Disposable
  • Designed for small spills of chemicals and hazardous liquids
  • Includes an autoclavable disposal bag that is heat resistant up to 145°C
  • For safe disposal of laboratory waste
Specifications
Color Green
Type Chemical Sorbent
Includes 2 pads (40 x 50 cm), 1 pair of disposable chemical protective gloves, 1 autoclavable disposal bag, 1 disposable brush, and multilingual instruction guide
Alternate Part Number SKH-MINI