Description
20G Balaclava Hood
- Material: 20G Spun Polypropylene
- Elastic Band at Eyes Opening
- 36 cm from Crown to Neck
- Full Head Enclosure
- Elastic Band at Neck
- Two-Piece Cutting
- PFAS Free
Description: Ideal for food handling, these colour-coded balaclava hoods are perfect for a wide range of hygienic work environments. The balaclava provides full head coverage and is designed to ensure a secure and comfortable fit with its elastic bands.
Greener Choice Claims
- Environmental benefits include:
- Recycled Content
Specifications
Specifications
Product Type | Protective Cap |
Size | One Size |
Material | Polypropylene |
Color | Red |
Closure Type | Ties |
Disposable | Single-use |
Latex Free | Yes |
Sterility | Non-sterile |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Chemsplash Disposable PolyProp Balaclava >
Spot an opportunity for improvement?Share a Content Correction