Learn More
B Braun™ Injekt™ Solo Single-use Syringe
Made of polypropylene and polyethylene B Braun™ Injekt™ Solo Single-use Syringe includes a green plunger with a highly transparent barrel. „Latex-free/PVC-free „„Silicone oil-free tube easily aspirates up to maximum volume.
Brand:  B Braun™ 9202528
Product Code. 10641200
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
This item is not returnable. View return policy
Description
Description
2-piece single-use syringe
  • Made of polypropylene/polyethylene
  • „„Green plunger
  • „„Highly transparent barrel „
  • „High contrast black graduation with permanent marking provides ideal readability „„
  • Safe plunger backstop
  • „„Easy aspiration up to maximum volume „„
  • Latex-free/PVC-free „„Silicone oil-free
Specifications
Specifications
Polypropylene, Polyethylene
3 mL
Centric Luer Cone
Single-use
Non-sterile
EN ISO 7886-1
Green Plunger
Yes