Learn More
Lenz Laborglasintrumente Distilling Links
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand: Lenz Laborglasintrumente 05040001
This item is not returnable.
View return policy
Description
- Distilling links without condenser
- With two vertical NS cones
- Made of DURAN™ tubing
Specifications
150 mm | |
Borosilicate Glass 3.3 | |
NS 14/23 |