missing translation for 'onlineSavingsMsg'
Learn More

Lenz Laborglasintrumente Distilling Links with Liebig Condenser and GL Thread

Product Code. 11562324
Click to view available options
Standard Taper Joint:
14/23 NS
29/32 NS
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11562324

Brand: Lenz Laborglasintrumente 05050414

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.

  • Distilling links with Liebig condenser
  • Outlet with NS cone, water connection with thread GL14
  • With socket NS 14/23 for distillation thermometers of 55 mm built-in length
  • Made of DURAN™ tubing

Specifications

Length (Metric) 400 mm
Material Duran Glass
Quantity 1 Ea.
Standard Taper Joint 29/32 NS
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.