missing translation for 'onlineSavingsMsg'
Learn More
DL-2-Bromopropionic acid, 99%, Thermo Scientific Chemicals Product Code.: 10192750

DL-2-Bromopropionic acid, 99%, Thermo Scientific Chemicals

Product Code. 10192750
1 kg, Glass bottle
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10192750

Brand: Thermo Scientific Acros 107350010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 598-72-1
Molecular Formula C3H5BrO2
Molecular Weight (g/mol) 152.98
MDL Number MFCD00004211,MFCD00145205
InChI Key MONMFXREYOKQTI-UHFFFAOYNA-N
Synonym 2-bromopropionic acid, propanoic acid, 2-bromo, dl-2-bromopropionic, propionic acid, 2-bromo, alpha-bromopropionic acid, dl-2-bromopropionic acid, 2-bromo-propionic acid, .alpha.-bromopropionic acid, propanoic acid, bromo, propanoicacid, 2-bromo
PubChem CID 11729
IUPAC Name 2-bromopropanoic acid
SMILES CC(Br)C(O)=O

Specifications

Color Colorless to Yellow
Density 1.6500g/mL
Boiling Point 203°C
Flash Point 100°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Linear Formula CH3CH(Br)CO2H
Refractive Index 1.4740 to 1.476
Quantity 1 kg
Beilstein 02, 254
Specific Gravity 1.65
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol and ether
Formula Weight 152.98
Percent Purity 99%
Physical Form Liquid After Melting
Chemical Name or Material DL-2-Bromopropionic acid
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0309061 Certificate TypeCertificate of Analysis Date03/01/2024 Catalog Number107350010,107352500,107350025
Lot NumberA0447430 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number107350010,107352500,107350025
Lot NumberA0431338 Certificate TypeCertificate of Analysis Date23/06/2021 Catalog Number107350010,107352500,107350025
compliance-icons compliance-icons
Product Identifier
  • DL-2-Bromopropionic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
DL-2-Bromopropionic acid, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.