missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-Aspartic Acid +99%, Thermo Scientific Chemicals
20.80 € - 124.00 €
Chemical Identifiers
CAS | 617-45-8 |
---|---|
Molecular Formula | C4H7NO4 |
Molecular Weight (g/mol) | 133.10 |
MDL Number | MFCD00063083 |
InChI Key | CKLJMWTZIZZHCS-UHFFFAOYNA-N |
Synonym | dl-aspartic acid, 2-aminosuccinic acid, dl-aminosuccinic acid, dl-asparagic acid, aspartic acid, dl, dl-asp-oh, +--aspartic acid, r,s-aspartic acid, h-dl-asp-oh, acid d,l-aspart |
PubChem CID | 424 |
ChEBI | CHEBI:22660 |
IUPAC Name | 2-aminobutanedioic acid |
SMILES | NC(CC(O)=O)C(O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10385970
|
Thermo Scientific Acros
105030050 |
5 g | Glass Bottle |
20.80 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10636672
|
Thermo Scientific Acros
105032500 |
250 g | Plastic Bottle |
46.60 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10721735
|
Thermo Scientific Acros
105030010 |
1 kg | Plastic Bottle |
124.00 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
617-45-8 | |
133.10 | |
CKLJMWTZIZZHCS-UHFFFAOYNA-N | |
424 | |
2-aminobutanedioic acid |
C4H7NO4 | |
MFCD00063083 | |
dl-aspartic acid, 2-aminosuccinic acid, dl-aminosuccinic acid, dl-asparagic acid, aspartic acid, dl, dl-asp-oh, +--aspartic acid, r,s-aspartic acid, h-dl-asp-oh, acid d,l-aspart | |
CHEBI:22660 | |
NC(CC(O)=O)C(O)=O |
Specifications
500ppm max. | |
Yellow | |
99+% | |
Authentic | |
10ppm max. | |
04, 483 | |
10, 853 | |
CKLJMWTZIZZHCS-UHFFFAOYNA-N | |
2-aminobutanedioic acid | |
133.10 | |
CHEBI:22660 | |
133.1 | |
99.0 to 101.0% (Acid) | |
Crystalline Low Melting Mass |
617-45-8 | |
10ppm max. | |
C4H7NO4 | |
HO2CCH2CH(NH2)CO2H | |
MFCD00063083 | |
0.3% max. (105°C, 3 hrs) | |
dl-aspartic acid, 2-aminosuccinic acid, dl-aminosuccinic acid, dl-asparagic acid, aspartic acid, dl, dl-asp-oh, +--aspartic acid, r,s-aspartic acid, h-dl-asp-oh, acid d,l-aspart | |
NC(CC(O)=O)C(O)=O | |
−0.3° to +0.3° (20°C, 589nm) (c=10, 2 N HCl) | |
424 | |
500ppm max. | |
95% min. (c=10, 2 N HCl, 430nm) 1cm cell | |
0.1% max. | |
DL-Aspartic acid, 99+% |
Safety and Handling
missing translation for 'einecsNumber' : 210-513-3
missing translation for 'rtecsNumber' : CI9097800
missing translation for 'tsca' : TSCA
RUO – Research Use Only