missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals DL-Cysteine, 97%

66.90 € - 220.00 €

Chemical Identifiers

CAS 3374-22-9
Molecular Formula C3H7NO2S
Molecular Weight (g/mol) 121.15
MDL Number MFCD00004881
InChI Key XUJNEKJLAYXESH-UHFFFAOYNA-N
Synonym dl-cysteine, cysteine, 2-amino-3-mercaptopropanoic acid, dl-cystein, cysteine, dl, +--cysteine, dl-cysteina polish, h-dl-cys-oh, 2-amino-3-mercaptopropionic acid, l-hsch2ch nh2 cooh
PubChem CID 594
ChEBI CHEBI:15356
IUPAC Name 2-amino-3-sulfanylpropanoic acid
SMILES NC(CS)C(O)=O
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
11320686
View Documents Promotion Details
Thermo Scientific Chemicals
230120100
10 g Glass Bottle
66.90 €
10g
Please to purchase this item. Need a web account? Register with us today!
11330686
View Documents Promotion Details
Thermo Scientific Chemicals
230120500
50 g Glass Bottle
220.00 €
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

3374-22-9
121.15
XUJNEKJLAYXESH-UHFFFAOYNA-N
594
2-amino-3-sulfanylpropanoic acid
C3H7NO2S
MFCD00004881
dl-cysteine, cysteine, 2-amino-3-mercaptopropanoic acid, dl-cystein, cysteine, dl, +--cysteine, dl-cysteina polish, h-dl-cys-oh, 2-amino-3-mercaptopropionic acid, l-hsch2ch nh2 cooh
CHEBI:15356
NC(CS)C(O)=O

Specifications

225.0°C
White
C3H7NO2S
HSCH2CH(NH2)CO2H
04, IV, 3145
dl-cysteine, cysteine, 2-amino-3-mercaptopropanoic acid, dl-cystein, cysteine, dl, +--cysteine, dl-cysteina polish, h-dl-cys-oh, 2-amino-3-mercaptopropionic acid, l-hsch2ch nh2 cooh
XUJNEKJLAYXESH-UHFFFAOYNA-N
2-amino-3-sulfanylpropanoic acid
594
121.15
Powder
3374-22-9
97%
Authentic
MFCD00004881
10, 2775
Solubility in water: soluble. Other solubilities: freely soluble in alcohol, insolublein ether, acetone, benzene, carbon, disulfide, ethyl acetate and carbon tetrachloride
NC(CS)C(O)=O
121.15
CHEBI:15356
97%
DL-Cysteine, 99%
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 222-160-2

missing translation for 'rtecsNumber' : HA1595000

Product Identifier
  • DL-Cysteine

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only