missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals DL-Methionine, 99+%
25.00 € - 81.30 €
Chemical Identifiers
CAS | 59-51-8 |
---|---|
Molecular Formula | C5H11NO2S |
Molecular Weight (g/mol) | 149.21 |
MDL Number | MFCD00063096 |
InChI Key | FFEARJCKVFRZRR-UHFFFAOYNA-N |
Synonym | dl-methionine, methionine, racemethionine, acimetion, banthionine, cynaron, lobamine, meonine, mertionin, metione |
PubChem CID | 876 |
ChEBI | CHEBI:16811 |
SMILES | CSCCC(N)C(O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10326960
|
Thermo Scientific Chemicals
125650025 |
2.5 g | Glass Bottle |
25.00 €
2.50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10457130
|
Thermo Scientific Chemicals
125651000 |
100 g | Plastic Bottle |
26.40 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10666132
|
Thermo Scientific Chemicals
125652500 |
250 g | Plastic Bottle |
40.30 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10510224
|
Thermo Scientific Chemicals
125650010 |
1 kg | Plastic Bottle |
81.30 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
59-51-8 | |
149.21 | |
FFEARJCKVFRZRR-UHFFFAOYNA-N | |
876 | |
CSCCC(N)C(O)=O |
C5H11NO2S | |
MFCD00063096 | |
dl-methionine, methionine, racemethionine, acimetion, banthionine, cynaron, lobamine, meonine, mertionin, metione | |
CHEBI:16811 |
Specifications
1000ppm max. | |
5.4 to 6.1 (2% w/v aq. soln.) | |
200ppm max. | |
99+% | |
Authentic | |
MFCD00063096 | |
0.5% max. (1 g, 105°C) | |
0.2% max. (TLC) | |
FFEARJCKVFRZRR-UHFFFAOYNA-N | |
2-amino-4-(methylsulfanyl)butanoic acid | |
876 | |
200ppm max. | |
95% min. (c=10, 2N HCl, 430nm) 1cm cell | |
0.1% max. | |
Crystalline Powder or Crystals |
59-51-8 | |
White | |
10ppm max. | |
C5H11NO2S | |
CH3SCH2CH2CH(NH2)CO2H | |
04, II, 938 | |
15, 6047 | |
dl-methionine, methionine, racemethionine, acimetion, banthionine, cynaron, lobamine, meonine, mertionin, metione | |
CSCCC(N)C(O)=O | |
149.21 | |
CHEBI:16811 | |
149.21 | |
99.0 to 101.0% (on dried substance) | |
1.34g/mL | |
DL-Methionine, 99+% |
Safety and Handling
missing translation for 'einecsNumber' : 200-432-1
missing translation for 'rtecsNumber' : PD0456000
missing translation for 'tsca' : TSCA
RUO – Research Use Only