missing translation for 'onlineSavingsMsg'
Learn More

DL-Serine methyl ester hydrochloride, 98+%, Thermo Scientific Chemicals

Product Code. 11411618
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5619-04-5
Molecular Formula C4H10ClNO3
Molecular Weight (g/mol) 155.58
MDL Number MFCD00012593
InChI Key NDBQJIBNNUJNHA-UHFFFAOYNA-N
Synonym dl-serine methyl ester hydrochloride, h-dl-ser-ome.hcl, methyl 2-amino-3-hydroxypropanoate hydrochloride, l-serine, hydrochloride, methyl dl-serinate hcl, d,l-serine methyl ester hydrochloride, zlchem 219, dl-serinemethylesterhydrochloride, h-ser-ome??hcl, pubchem10908
PubChem CID 517373
IUPAC Name methyl 2-amino-3-hydroxypropanoate;hydrochloride
SMILES COC(=O)C(CO)N.Cl

Specifications

Melting Point ∼134°C (decomposition)
Beilstein 6067970
Formula Weight 155.58
Percent Purity ≥98%
Quantity 250 g
Sensitivity Moisture sensitive
Chemical Name or Material DL-Serine methyl ester hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'