missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals DL-Tyrosine, 98%

Product Code. 11414774
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

The tyrosine of this diet was labeled by adding to it an amount of synthetic, isotopic dl-tyrosine cor- responding to twice that already present in the food. Incorporation of total C14 into muscle protein was reduced in 186-day infected mice fed dl-tyrosine-2-C-14 as compared to muscle proteins obtained from noninfected animals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The tyrosine of this diet was labeled by adding to it an amount of synthetic, isotopic dl-tyrosine cor- responding to twice that already present in the food. Incorporation of total C14 into muscle protein was reduced in 186-day infected mice fed dl-tyrosine-2-C-14 as compared to muscle proteins obtained from noninfected animals.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 556-03-6
Molecular Formula C9H11NO3
Molecular Weight (g/mol) 181.19
MDL Number MFCD00063074
InChI Key OUYCCCASQSFEME-UHFFFAOYNA-N
Synonym dl-tyrosine, h-dl-tyr-oh, 2-amino-3-4-hydroxyphenyl propanoic acid, tyrosin, tyrosine, dl, l-tyrosine, free base, tirosina, l-tryosine, 3-4-hydroxyphenyl-dl-alanine, benzenepropanoic acid, s
PubChem CID 1153
ChEBI CHEBI:18186
IUPAC Name 2-amino-3-(4-hydroxyphenyl)propanoic acid
SMILES NC(CC1=CC=C(O)C=C1)C(O)=O

Specifications

Melting Point ∼325°C (decomposition)
Beilstein 515881
Merck Index 14,9839
Solubility Information Soluble in water.
Formula Weight 181.19
Percent Purity 98%
Quantity 250 g
Chemical Name or Material DL-Tyrosine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'