missing translation for 'onlineSavingsMsg'
Learn More
Double Distilled Water, Carl Roth™ Product Code.: 10628013
Add to basket

Double Distilled Water, Carl Roth™

Product Code. 10628013
1 L
Encompass
Click to view available options
Quantity:
1 L
This item is not returnable. View return policy

Product Code. 10628013

Brand: Carl Roth 3478.2

Add to basket
Add to basket
This item is not returnable. View return policy

Redistilled. Roth Carl™ Double Distilled Water can be used for a wide range of laboratory purposes.

  • Redistilled
  • Empirical formula H2O
  • Molar mass (M) 18,02 g/mol
  • Density (D) 1,0 
  • Boiling point (bp) 100 °C
  • Melting point (mp)0 °C
  • CAS-Nr. [7732-18-5]
  • EG-Nr. 231-791-2

Specifications

Quantity 1 L
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
Double Distilled Water, Carl Roth™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.