Learn More
Dräger™ CMS Chips: Carbon Dioxide
Click to view available options
Measurement Range:
1 to 20%
1000 to 25000
200 to 3000
Response Time:
12 to 120 sec.
25 to 140 sec.
60 to 260 sec.
Unit Size:
Each
Description
Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.
Specifications
Specifications
| Air Pressure | 700 to 1100 hpa |
| Type | Carbon Dioxide CMS Chip |
| Humidity Range | 1 to 30mg/L |
| Measurement Range | 200 to 3000 |
| Response Time | 60 to 260 sec. |
| Temperature Range (English) | 32°F to 104°F |
| Temperature Range (Metric) | 0°C to 40°C |