Learn More
Dräger™ CMS Chips: Petroleum Hydrocarbons
Click to view available options
Measurement Range:
100 to 3000
20 to 500
Response Time:
150 to 330 sec.
30 to 110 sec.
Unit Size:
Each
Description
Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.
Specifications
Specifications
| Air Pressure | 700 to 1100 hpa |
| Type | Petroleum Hydrocarbons CMS Chip |
| Humidity Range | 1 to 30mg/L |
| Measurement Range | 20 to 500 |
| Response Time | 150 to 330 sec. |
| Temperature Range (English) | 32°F to 104°F |
| Temperature Range (Metric) | 0°C to 40°C |