Learn More
Krüss Optronic™ DR201-95 Model Digital Hand-Held Refractometer
Carry out random refractive index checks or regulate mixing ratios easily with this mobile, hand-held refractometer. Krüss Optronic™ DR201-95 Model Digital Hand-Held Refractometer saves time and effort by facilitating on-site measuring.
Brand:  Krüss Optronic™ DR201-95
Product Code. 10277322
  • 763.00 € / Each
Estimated Shipment: 12-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • Automatic measurement
  • High measuring accuracy
  • Fast to fill and clean
  • Sample tray made of stainless steel
  • Robust housing and low weight
  • Automatic temperature compensation (Brix scale)
  • Display of the measurement results in different units
  • Very simple operation
Specifications
Specifications
Handheld Digital Refractometer
1.3330 to 1.5318
1.3330 to 1.5318 nD/0.2% Brix
Determination of mixing ratios, quality and quantity inspection
130 x 80 x 40 mm
10°C to 40°C
1.5 V Battery
0 to 95 Brix
DR201-95
0.0001nD/0.1 % Brix
200 g
0.5°C
0°C to 40°C