missing translation for 'onlineSavingsMsg'
Learn More

DSP-4, Tocris Bioscience™

Product Code. 18065482
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Adrenergic neurotoxin

Adrenergic neurotoxin. Displays neurotoxic effects on both peripheral and central noradrenergic neurons.

Chemical Identifiers

CAS 40616-75-9
Molecular Formula C11H16BrCl2N
Molecular Weight (g/mol) 313.06
InChI Key NDDRNRRNYOULND-UHFFFAOYSA-N
Synonym dsp-4 hydrochloride, dsp-4, n-2-chloroethyl-n-ethyl-2-bromobenzylamine hydrochloride, n-ethyl-n-2-chloroethyl-2-bromobenzylamine hydrochloride, n-2-bromobenzyl-2-chloro-n-ethylethanamine hydrochloride, 2-bromophenyl methyl 2-chloroethyl ethylamine hydrochloride, n-2-chloroethyl-n-ethyl-2-bromobenzylamine hydrochloride, solid
PubChem CID 11957497
IUPAC Name N-[(2-bromophenyl)methyl]-2-chloro-N-ethylethanamine;hydrochloride
SMILES CCN(CCCl)CC1=CC=CC=C1Br.Cl

Specifications

Quantity 10 mg
Formula Weight 313.06
Percent Purity >99%
Chemical Name or Material DSP-4
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'