missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuraCal™ 5.00 Value pH Buffer, Hamilton™
Click to view available options
Quantity:
500 mL
Description
- 5.00 pH value
- Immune to micro-organisms
- Expiration date on the bottle
- First class certificate with traceability to international standards
Specifications
Specifications
pH | 5.00 |
Physical Form | Liquid |
Quantity | 500 mL |
Solution Type | Buffer |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'