Learn More
Lenz Laborglasinstrumente™ Duran™ Glass Socket Adapter
Click to view available options
Cone Size:
NS 29/32
NS 45/40
Unit Size:
Pack of 10
Specifications
Specifications
| Certifications/Compliance | DIN 12249 form E and in addition to DIN standard |
| Cone Size | NS 45/40 |
| Length (Metric) | 145mm |
| Material | Duran Glass |
| Product Type | Socket Adapter |
| Quantity | 10Pack |