missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences DURAN™ Separating Funnel stopcock Gilson, complete with PTFE key

For attaching to DURAN™ separating funnel raw bodies.

Brand:  DWK Life Sciences 286430001

1216.00 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 15884738

  • 1280.00 € / Pack of 25
Estimated Shipment: 02-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

DURAN™ Gilson separating funnel taps are components for the production of Gilson separating funnels with a particularly strong approach to reducing the risk of breakage. In the polished version with PTFE plug suitable for grease free working.

  • Separating funnel tap sleeve with PTFE plug
  • Fat-free working through the polished tap surface
  • With a particularly strong approach to reducing the risk of breakage
  • Glass articles for further processing
TRUSTED_SUSTAINABILITY
Specifications

Specifications

4 mm
Borosilicate Glass (3.3), PTFE
18.8 NS
25/Pk.
24 mm
Autoclavable
1.8 mm
For attachment to DURAN™ separating funnel raw bodies; For the production of separating funnels according to Gilson
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions