Learn More
Abeba™ Dynamic 37312 Shoes
ESD dissipative shoe includes special respiration absorbing lining for the foot to breathe.
66.43 €
Specifications
Certifications/Compliance | CE EN ISO 20347:2012 OB A, E, FO SRC |
---|---|
Color | White |
Material | Leather |
Sole Features | Slip-resistant |
Sole Material | Polyurethane |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
15660848
|
Abeba™
37312/35 |
35 |
66.43 €
1 pair |
Estimated Shipment: 04-10-2024
Log in to see stock. |
|||||
15670848
|
Abeba™
37312/36 |
36 |
66.43 €
1 pair |
Estimated Shipment: 04-10-2024
Log in to see stock. |
|||||
15680848
|
Abeba™
37312/37 |
37 |
66.43 €
1 pair |
Estimated Shipment: 04-10-2024
Log in to see stock. |
|||||
15690848
|
Abeba™
37312/38 |
38 |
66.43 €
1 pair |
Estimated Shipment: 27-09-2024
Log in to see stock. |
|||||
15600858
|
Abeba™
37312/39 |
39 |
66.43 €
1 pair |
Estimated Shipment: 27-09-2024
Log in to see stock. |
|||||
15610858
|
Abeba™
37312/40 |
40 |
66.43 €
1 pair |
Estimated Shipment: 27-09-2024
Log in to see stock. |
|||||
15620858
|
Abeba™
37312/41 |
41 |
66.43 €
1 pair |
Estimated Shipment: 27-09-2024
Log in to see stock. |
|||||
15630858
|
Abeba™
37312/42 |
42 |
66.43 €
1 pair |
Estimated Shipment: 04-10-2024
Log in to see stock. |
|||||
15640858
|
Abeba™
37312/43 |
43 |
66.43 €
1 pair |
Estimated Shipment: 04-10-2024
Log in to see stock. |
|||||
15650858
|
Abeba™
37312/44 |
44 |
66.43 €
1 pair |
Estimated Shipment: 04-10-2024
Log in to see stock. |
|||||
15660858
|
Abeba™
37312/45 |
45 |
66.43 €
1 pair |
Estimated Shipment: 04-10-2024
Log in to see stock. |
|||||
15670858
|
Abeba™
37312/46 |
46 |
66.43 €
1 pair |
Estimated Shipment: 27-09-2024
Log in to see stock. |
|||||
15680858
|
Abeba™
37312/47 |
47 |
66.43 €
1 pair |
Estimated Shipment: 04-10-2024
Log in to see stock. |
|||||
15690858
|
Abeba™
37312/48 |
48 |
66.43 €
1 pair |
Estimated Shipment: 04-10-2024
Log in to see stock. |
|||||
Description
- CE EN ISO 20347:2012 OB A, E, FO SRC
- Smooth leather white/blue
- Slip resistance TPU outsole sole
- Replaceable acc Wave insole (article no. 3574)
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Heel strap foldable, adjustable
Specifications
CE EN ISO 20347:2012 OB A, E, FO SRC | |
Leather | |
Polyurethane |
White | |
Slip-resistant | |
37312 |