missing translation for 'onlineSavingsMsg'
Learn More

Dysprosium foil, 0.25mm (0.01 in.) thick, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11381769
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7429-91-6
Molecular Formula Dy
Molecular Weight (g/mol) 162.50
MDL Number MFCD00010982
InChI Key KBQHZAAAGSGFKK-UHFFFAOYSA-N
Synonym ingot reo, disprosio, atom, ion, ingot, pieces, powder, foil, 3n, chips, 3n, ingot, 3n
PubChem CID 23912
ChEBI CHEBI:33377
IUPAC Name dysprosium
SMILES [Dy]

Specifications

UN Number UN3178
Quantity 25 x 25 mm
Percent Purity 99.9%
Odor Odorless
Weight ≈1.33g/25x25mm
Form Foil
Assay (REO)
Chemical Name or Material Dysprosium foil, 0.25mm (0.01 in.) thick

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'