Learn More
3M™ E-A-R™ Classic™ Small Earplugs
The diameter of E-A-R™ Classic™ Small Earplug is 7% less than the Classic earplug (and the same length) to help provide optimum comfort and protection in smaller ears.
Brand: 3M™ 7000089396
356.16 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
- The E-A-R™ Classic™ Small Earplugs may be used for protection against moderate to high noise environments, providing an SNR of 29 dB as fully tested and approved against EN 352-2:2020.
- Uncorded
Specifications
Small | |
Uncorded | |
Single-use | |
No |
29 dB | |
Cylinder | |
No | |
No |
Product Suggestions
Customers who viewed this item also viewed
Promotions
-
Multiple Manufacturers
Gear Up for Safety
Save up to 60% on gloves, apparel, and morePromo Code: 24131Expires: 2025-06-08
New Offer Shop Now
Your input is important to us. Please complete this form to provide feedback related to the content on this product.