Learn More
STEMCELL Technologies EasySep™ HLA Chimerism Whole Blood CD15 Positive Selection Kit
Immunomagnetic positive selection of human CD15+ cells (e.g. myeloid cells) from whole blood
Brand:  STEMCELL Technologies 17881
Encompass
Product Code. 17137791
  • 1185.00 € / 1 set
Estimated Shipment: 19-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • The EasySep™ HLA Chimerism Whole Blood CD15 Positive Selection Kit is designed to isolate highly purified CD15+ cells from fresh whole blood by positive selection
  • Desired cells are targeted with Tetrameric Antibody Complexes recognizing CD15 and dextran-coated magnetic particles
  • Labeled cells are separated using an EasySep™ magnet without the use of columns
  • Cells of interest remain in the tube while unwanted cells are poured off
  • The CD15 antigen is expressed on granulocytes and monocytes
  • This product replaces the EasySep™ Human Whole Blood CD15 Positive Selection Kit (Catalog #18681) and EasySep™ HLA Whole Blood CD15 Positive Selection Kit (Catalog #18681HLA), for even faster cell isolations
Specifications
Specifications
Myeloid Cells
Cell Isolation
60 mL Buffy Coat or Whole Blood