Learn More
STEMCELL Technologies EasySep™ HLA Chimerism Whole Blood CD15 Positive Selection Kit
Immunomagnetic positive selection of human CD15+ cells (e.g. myeloid cells) from whole blood
Brand: STEMCELL Technologies 17881
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
This item is not returnable.
View return policy
Description
- The EasySep™ HLA Chimerism Whole Blood CD15 Positive Selection Kit is designed to isolate highly purified CD15+ cells from fresh whole blood by positive selection
- Desired cells are targeted with Tetrameric Antibody Complexes recognizing CD15 and dextran-coated magnetic particles
- Labeled cells are separated using an EasySep™ magnet without the use of columns
- Cells of interest remain in the tube while unwanted cells are poured off
- The CD15 antigen is expressed on granulocytes and monocytes
- This product replaces the EasySep™ Human Whole Blood CD15 Positive Selection Kit (Catalog #18681) and EasySep™ HLA Whole Blood CD15 Positive Selection Kit (Catalog #18681HLA), for even faster cell isolations
Specifications
Myeloid Cells | |
Cell Isolation | |
60 mL Buffy Coat or Whole Blood |