missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies EasySep™ Human CD4+CD127lowCD25+ Regulatory T Cell Isolation Kit
missing translation for 'orderingAttributeHoverText'
Quantity:
1 x 109 cells
missing translation for 'unitSize'
Each
Description
- This kit isolates human CD4+CD127lowCD25+ regulatory T cells (Tregs) from fresh peripheral blood mononuclear cells (PBMCs) or leukapheresis samples
- First, CD25+ cells are isolated by column-free immunomagnetic positive selection using EasySep™ Releasable RapidSpheres™
- Then, bound magnetic particles are removed from the EasySep™-isolated CD25+ cells, and unwanted non-Tregs are targeted for depletion
- The final isolated fraction contains highly purified CD4+CD127lowCD25+ cells that express high levels of FOXP3 and are immediately ready for downstream applications
- An optional protocol allows for the isolation of CD4+CD25- responder T cells in parallel for use in functional studies
- Following cell isolation with this EasySep™ kit, antibody complexes remain bound to the cell surface and may interact with Brilliant Violet™ antibody conjugates, polyethylene glycol-modified proteins, or other chemically related ligands
Specifications
Specifications
| Cell Type | T Cells, T Cells, CD4+, T Cells, Regulatory |
| For Use With (Application) | Cell Isolation |
| Quantity | 1 x 109 cells |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction