Learn More
R&DSystems™ EAV NSP9 (NP_705590.1) VersaClone cDNA
Contains gene inserts that are fully sequenced and produced with high quality
Brand:  R&DSystems™ RDC3443
Product Code. 18461430
  • 309.00 € / 10µg
Please to purchase this item. Need a web account? Register with us today!
Explore more special offers
This item is not returnable. View return policy
Description
Description
TRUSTED_SUSTAINABILITY
Specifications
Specifications
nsp9
1489749
Store the unopened product at -20 to -70 °C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
10 μg
NP_705590.1
Non-structural protein 9, ORF1a polyprotein, ORF1ab polyprotein, p12
EAV
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
R&DSystems™ EAV NSP9 (NP_705590.1) VersaClone cDNA > 10 μg