Learn More
Elma™ EC 10 Cleaner
Providing powerful cleaning power for through ultrasonic and immersion cleaning of alkali-resistant parts and medical devices made of stainless steel, steel, veramic, glass, plastic and rubber.
Brand:  Elma™ 5804400000
Product Code. 10467411
  • 300.00 € / 25 L
Please to purchase this item. Need a web account? Register with us today!
Explore more special offers
This item is not returnable. View return policy
Description
Description
  • Suitable for cleaning of alkali-resistant parts and medical devices such as medical and dental instruments, crowns, impression trays, laboratory equipment etc. made of stainless steel, steel, veramic, glass, plastic and rubber.
  • Removes blood, salica, proteins, tissue and bone residues as well as fats, oils, grinding and polishing pastes.
  • Not suitable for aluminum, zinc and light metal alloys as well as for alkali-sensitive plastics and plastics that are susceptible for stress cracking e.g. ABS and PC.
Specifications
Specifications
EC 10 Cleaner
CE
For the thorough ultrasonic cleaning of medical and dental instruments and laboratory equipments