missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EFTUD2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00 € - 410.00 €
Specifications
| Antigen | EFTUD2 |
|---|---|
| Concentration | 0.23 mg/mL |
| Applications | Immunocytochemistry, Western Blot, Immunofluorescence, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16836443
|
Proteintech
10208-1-AP-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16826443
|
Proteintech
10208-1-AP-150UL |
150 μL |
410.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
10208-1-AP is a rabbit polyclonal antibody raised against the N-terminal of human EFTUD2.
EFTUD2 is component of the U5 snRNP complex required for pre-mRNA splicing.Specifications
| EFTUD2 | |
| Immunocytochemistry, Western Blot, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Rat, Mouse, Human | |
| O08810, Q15029 | |
| 20624, 287739, 9343 | |
| EFTUD2 Fusion Protein Ag0279 | |
| Primary | |
| -20°C | |
| EFTUD2 |
| 0.23 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| EFTUD2, hSNU114, KIAA0031, Snrp116, Snu114, SNU114 homolog, U5 116 kDa, U5 116KD | |
| Eftud2 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title