missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EGR1/EGR2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA538588
This item is not returnable.
View return policy
Description
The protein encoded by this gene belongs to the EGR family of C2H2-type zinc-finger proteins. It is a nuclear protein and functions as a transcriptional regulator. The products of target genes it activates are required for differentitation and mitogenesis. Studies suggest this is a cancer suppresor gene.
Specifications
| EGR1/EGR2 | |
| Polyclonal | |
| Unconjugated | |
| EGR1 | |
| TIS8, AT225, G0S30, NGFI-A, ZNF225, KROX-24, ZIF-268, EGR1, EGR2; early growth response 2 (Krox-20 homolog, Drosophila) ; CMT1D; CMT4E; DKFZp686J1957; FLJ14547; KROX20; KROX-20, Drosophila, homolog early growth response-2; Krox-20 homolog | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 114090, 13653, 13654, 1958, 1959, 24330 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P11161, P18146, P08046, P08152, P08154, P51774 | |
| EGR1, EGR2 | |
| A synthetic peptide derived from the internal region of human EGR1/2 | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction