missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ ELAVL1 293T Cell Transient Overexpression Lysate (Denatured) (T03)
Human overexpression lysate
Brand: Abnova™ H00001994-T03.100uL
Additional Details : Weight : 0.00010kg
Specifications
Western Blot | |
ELAVL1 293T Cell Transient Overexpression Lysate(Denatured) | |
Theoretical M.W. 36.1kDa | |
Transient overexpression cell lysate was tested with anti-ELAVL1 antibody (Mfr. No. H00001994-D01) by Western Blot. | |
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
293T | |
BC003376 | |
pCMV-ELAVL1 full-length | |
100 μL | |
Human |