missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Knick Elektronische™ 766 Laboratory pH Meter with Combination Electrode
Includes model 766 ph Meter with ph Sensor, attachable stand and buffer set.
Brand: Knick Elektronische™ 766-SETB
This item is not returnable.
View return policy
Description
- Gaincheck automatic device test
- Trueline calibrated analog recorder output
- Sensor monitoring with Sensoface icon alerts
- Automatic calibration with patented Calimatic
- EMC according to NAMUR
- Simultaneous pH and temperature display
- Simple operation
- Leak-proof membrane keyboard
- Robust housing
- Protection category IP 54
- 3-year warranty
Specifications
Glass-reinforced polyamide 12, stainless steel cover, with connecting ZU 6954 attachable stand and buffer set | |
<0.3 K | |
<0.01 | |
0.01 | |
0.01 | |
50°C to 150°C | |
DIN 19262 | |
255 x 244 x 95 mm | |
Pt 1000, automatic selection, Manual: 0.0 to +100.0°C |
SE 100N ph Sensor | |
<0.1% ±0.3 | |
-2 to 16 | |
-1999 to 1999 | |
1, 2 or 3 | |
LED | |
IP-54 | |
230 V –15 % +10 %, 48 to 62 Hz, <10 VA, Option 363: 115 V AC |