Learn More
Merck MultiScreen™HTS 8-Well Strips with Immobilon™-P PVDF Membrane
Shop All Merck Products
Click to view available options
Product Type:
Microplate 8-Well Strip
Microplate Strip Support Frame
Unit Size:
Pack of 10
Description
Includes
Twelve 8-well strips in a support frame. Additional support frames also available separately.
- For ELISpot and protein binding assays
Specifications
Specifications
| Format | Strip Plate |
| Array | 1 x 8 |
| Color | Clear |
| Product Type | Microplate Strip Support Frame |
| For Use With (Equipment) | 8-well strips |
| Material | ABS Plastic |
| No. of Wells | 8 |
| Sterility | Sterile |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Condividi una correzione di contenuto