missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Endo Agar

Isolate and differentiate Enterobacteriaceae from potable water, dairy products and foods with this ready-to-use medium.

Brand:  Thermo Scientific™ PO5005A

Product Code. 10057022

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Isolate and differentiate Enterobacteriaceae from clinical samples, potable water, dairy products and foods1, 2 with Thermo Scientific™ Oxoid™ Endo Agar. Originally designed for the isolation of typhoid bacillus3, Endo Agar is now mostly used for the differentiation of lactose fermenting and lactose non-fermenting intestinal organisms.

Isolate and differentiate Enterobacteriaceae with Endo Agar.

  • Easy differentiation of lactose fermenting and non-lactose fermenting intestinal organisms.
  • Ready-to-use convenience of prepared media.

Coliforms are commonly defined by their ability to ferment lactose rapidly, producing acid and gas, typically within 24 hours. The genera that would be commonly expected to belong to this group include Enterobacter, Klebsiella, Citrobacter and Escherichia, particularly Escherichia coli. However, species belonging to other genera, for example Erwinia andSerratia, can also ferment lactose, albeit slowly, and some strains of Citrobacter and Klebsiella, as well as Salmonella arizonae and Hafnia alvei, show delayed or variable fermentation ability.

Production of acid and aldehyde by lactose fermenting organisms, such as Escherichia coli, gives rise to the characteristic red coloration of the colony and the surrounding medium.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Light pink to beige red
90mm Monoplate
Agar
10
2°C to 10°C
Germany
17g ± 5%
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number3316773 Certificate TypeCertificate of Analysis Date02/07/2021 Catalog NumberPO5005A
Lot Number3316772 Certificate TypeCertificate of Analysis Date02/07/2021 Catalog NumberPO5005A
Lot Number3316771 Certificate TypeCertificate of Analysis Date02/07/2021 Catalog NumberPO5005A
Lot Number3312480 Certificate TypeCertificate of Analysis Date02/07/2021 Catalog NumberPO5005A
Lot Number3309310 Certificate TypeCertificate of Analysis Date17/06/2021 Catalog NumberPO5005A
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Endo Agar >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.