Learn More
Memmert™ Entry Port 100 mm for UF750
Optional entry port for Memmert Ovens.
Brand: Memmert™ UF750F9
This item is not returnable.
View return policy
Description
- Entry port with 100 mm diameter
- Flap closure
- Custom position TBC in the back
Specifications
UF750 Oven | |
Entry Port | |
100 mm |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Memmert™ Entry Port 100 mm for UF750 > For Use With: UF750
Spot an opportunity for improvement?Share a Content Correction