Learn More
Memmert™ Entry Port 14 mm for UF160plus
Optional entry port for Memmert Ovens.
Brand:  Memmert™ UF160plusD6
Product Code. 17589065
  • 221.00 € / Each
Estimated Shipment: 26-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Entry port with 14 mm diameter
  • Flap closure
  • Custom position TBC in the back
Specifications
Specifications
UF160plus Oven
Entry Port
14 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Memmert™ Entry Port 14 mm for UF160plus > For Use With: UF160plus