Learn More
Memmert™ Entry Port 23 mm for UF1060, Custom TBC Back Side
Optional entry port for Memmert Ovens.
Brand: Memmert™ UF1060F6
This item is not returnable.
View return policy
Description
- Entry port with 23 mm diameter
- Flap closure
- Custom position TBC in the back
Specifications
UF1060 Oven | |
Entry Port | |
23 mm |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Memmert™ Entry Port 23 mm for UF1060, Custom TBC Back Side > For Use With: UF1060
Spot an opportunity for improvement?Share a Content Correction