Learn More
Memmert™ Entry Port 23 mm for UF160plus, Custom TBC Back Side
Optional entry port for Memmert Ovens.
Brand:  Memmert™ UF160plusF6
Product Code. 17529085
  • 195.00 € / Each
Estimated Shipment: 23-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Entry port with 23 mm diameter
  • Flap closure
  • Custom position TBC in the back
Specifications
Specifications
UF160plus Oven
Entry Port
23 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Memmert™ Entry Port 23 mm for UF160plus, Custom TBC Back Side > For Use With: UF160plus