Learn More
Memmert™ Entry Port 23 mm for UF75, Standard Position Left Side Center/Center
Optional entry port for Memmert Ovens.
Brand: Memmert™ UF75F0
This item is not returnable.
View return policy
Description
- Entry port with 23 mm diameter
- Flap and stopper closure
- Standard position left side center/center
Specifications
UF75 Oven | |
Entry Port | |
23 mm |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Memmert™ Entry Port 23 mm for UF75, Standard Position Left Side Center/Center > For Use With: UF75
Spot an opportunity for improvement?Share a Content Correction