Learn More
Memmert™ Entry Port 23 mm for UN160plus, Standard Position Right Side Center/Center
Product Code. 17539925 Shop All Memmert Products
Click to view available options
Diameter (Metric):
23 mm
Unit Size:
Each
This item is not returnable. View return policy
Product Code. 17539925
Brand: Memmert™ UN160plusF2
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
Optional entry port for Memmert Ovens.
  • Entry port with 23 mm diameter
  • Flap and stopper closure
  • Standard position right side center/center
Specifications
For Use With (Equipment) UN160plus Oven
Type Entry Port
Diameter (Metric) 23 mm