Learn More
Memmert™ Entry Port 38 mm for UN260plus
Product Code. 17500036 Shop All Memmert Products
Click to view available options
Diameter (Metric):
38 mm
Unit Size:
Each
This item is not returnable. View return policy
Product Code. 17500036
Brand: Memmert™ UN260plusF7
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
Optional entry port for Memmert Ovens.
  • Entry port with 38 mm clear diameter
  • Flap and stopper closure
  • Custom position TBC in the back
Specifications
For Use With (Equipment) UN260plus Oven
Type Entry Port
Diameter (Metric) 38 mm