Learn More
Memmert™ Entry Port 57 mm for UN160plus
Optional entry port for Memmert Ovens.
Brand: Memmert™ UN160plusF8
This item is not returnable.
View return policy
Description
- Entry port with 57 mm diameter
- Flap closure
- Custom position TBC in the back
Specifications
UN160plus Oven | |
Entry Port | |
57 mm |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Memmert™ Entry Port 57 mm for UN160plus > For Use With: UN160plus
Spot an opportunity for improvement?Share a Content Correction