Learn More
BRAND™ EPDM Rubber Conical Gaskets
Product Code. p-8003805 Shop All Brand Products
Click to view available options
Diameter (Metric) Outside:
11 mm
16 mm
20 mm
22 to 84 mm
25 mm
32 mm
43 mm
71 mm
89 mm
Unit Size:
Pack of 10
Pack of 8
Product Code. 10548781
Brand: BRAND™ 462512
Please to purchase this item. Need a web account? Register with us today!
Place between filter funnel and filter flask for a secure fit.
  • Rubber (EPDM) construction
  • Conical shape
  • Provides a secure fit between filter funnel and filter flask
  • Supplied in packs of 10
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Used for Filter Funnels and Filter Flasks
Length (Metric) 22 mm
Quantity 10
Diameter (Metric) Outside 11 mm
Material EPDM
Type Gasket, Conical