missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ESR1 Mouse anti-Canine, Human, Mouse, Clone: OTI3G2, TrueMAB™, Origene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA807239S
This item is not returnable.
View return policy
Description
ESR1 Monoclonal antibody specifically detects ESR1 in Canine, Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
ESR1 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P03372, P19785 | |
Mouse | |
Affinity chromatography | |
RUO | |
13982, 2099, 403640 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
OTI3G2 | |
Unconjugated | |
ESR1 | |
DKFZp686N23123; ER; Er alpha; ER36; ERa; ERalpha; ER-alpha; ERalpha protein; ESR; esr 1; Esr1; esr-1; ESRA; Estr; Estra; Estradiol receptor; estrogen nuclear receptor alpha; Estrogen receptor; estrogen receptor 1; estrogen receptor 1 (alpha); estrogen receptor alpha; estrogen receptor alpha c-terminus splice variant 1-2; estrogen receptor alpha E1-E2-1-2; estrogen receptor alpha E1-N2-E2-1-2; estrogen receptor alpha splice variant, CTERP-1; estrogen receptor alpha splice variant, ERalphaDup5; estrogen receptor alpha splice variant, ERalphai45a; estrogen receptor alpha splice variant, ERalphai45bL; estrogen receptor alpha splice variant, ERalphai45bS; estrogen receptor alpha splice variant, ERalphai45c; estrogen receptor alpha splice variant, ERalphai56; estrogen receptor alpha splice variant, ERalphai67; estrogen receptor alpha variant delta 4; estrogen receptor protein; estrogen receptor, alpha; ESTRR; hER-alpha36; I79_001166; Nr3a1; nuclear receptor; nuclear receptor subfamily 3 group A member 1; RNESTROR; RP1-130E4.1; unnamed protein product | |
Human recombinant protein fragment corresponding to amino acids 1-310 of human ESR1 produced in E.coli. | |
30 μL | |
Primary | |
Canine, Human, Mouse | |
Antibody | |
IgG2a |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction