Learn More
Estradiol valerianate, MedChemExpress
MedChemExpress Estradiol valerianate (β-estradiol 17-valerate) is a synthetic estrogen widely used in combination with other steroid hormones in hormone replacement therapy drugs.
Brand: Medchem Express HY-B0672

Description
Estradiol valerianate (β-estradiol 17-valerate) is a synthetic estrogen widely used in combination with other steroid hormones in hormone replacement therapy drugs.Specifications
Estradiol valerianate | |
White | |
C23H32O3 | |
β-Estradiol 17-valerate | |
C[C@@]1([C@H]2OC(CCCC)=O)[C@](CC2)([H])[C@@](CCC3=C4C=CC(O)=C3)([H])[C@]4([H])CC1 | |
356.5 | |
Research | |
Cancer-programmed cell death |
979-32-8 | |
Research | |
1 g | |
DMSO : ≥ 50 mg/mL (140.25 mM) ∣H2O : < 0.1 mg/mL (insoluble) | |
356.5 | |
99.24% | |
Solid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Research purposes only