missing translation for 'onlineSavingsMsg'
Learn More

Estradiol valerianate, MedChemExpress

Product Code. p-200097720
Encompass_Preferred
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 16441038

Brand: Medchem Express HYB0672

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Estradiol valerianate (β-estradiol 17-valerate) is a synthetic estrogen widely used in combination with other steroid hormones in hormone replacement therapy drugs.

Estradiol valerianate (β-estradiol 17-valerate) is a synthetic estrogen widely used in combination with other steroid hormones in hormone replacement therapy drugs.

Specifications

Chemical Name or Material Estradiol valerianate
CAS 979-32-8
Color White
Purity Grade Notes Research
Molecular Formula C23H32O3
Quantity 5 g
Synonym β-Estradiol 17-valerate
Solubility Information DMSO : ≥ 50 mg/mL (140.25 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES C[C@@]1([C@H]2OC(CCCC)=O)[C@](CC2)([H])[C@@](CCC3=C4C=CC(O)=C3)([H])[C@]4([H])CC1
Molecular Weight (g/mol) 356.5
Formula Weight 356.5
Percent Purity 99.24%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.