Learn More
Estramustine phosphate sodium, MedChemExpress
MedChemExpress Estramustine phosphate sodium, an estradiol analog, is an orally active antimicrotubule chemotherapy agent. Estramustine phosphate sodium depolymerises microtubules by binding to microtubule associated proteins (MAPs) and/or to tubulin. Estramustine phosphate sodium can interfere mitosis, trigger cell death and induce apoptosis, which can be used for the research of cancer like prostate cancer.
Brand:  Medchem Express HY-13627
Encompass_Preferred
 View more versions of this product
Product Code. 16426256
  • 195.75 € / 100mg
Please to purchase this item. Need a web account? Register with us today!
Explore more special offers
Description
Description
Estramustine phosphate sodium, an estradiol analog, is an orally active antimicrotubule chemotherapy agent. Estramustine phosphate sodium depolymerises microtubules by binding to microtubule associated proteins (MAPs) and/or to tubulin. Estramustine phosphate sodium can interfere mitosis, trigger cell death and induce apoptosis, which can be used for the research of cancer like prostate cancer.
Specifications
Specifications
Estramustine phosphate sodium
White
C23H30Cl2NNa2O6P
H2O : 62.5 mg/mL (110.75 mM; Need ultrasonic) ∣DMSO : 5 mg/mL (8.86 mM; Need ultrasonic)
564.35
96.79%
Solid
52205-73-9
Research
100 mg
O=C(N(CCCl)CCCl)OC1=CC2=C(C=C1)[C@@]3([H])CC[C@@]4(C)[C@](CC[C@@H]4OP(O[Na])(O[Na])=O)([H])[C@]3([H])CC2
564.35
Research
Cancer-programmed cell death