Learn More
Ethyl 2-cyano-2-methylpropionate, 97%, Thermo Scientific Chemicals
42.21 € - 140.70 €
Chemical Identifiers
CAS | 1572-98-1 |
---|---|
Molecular Formula | C7H11NO2 |
Molecular Weight (g/mol) | 141.17 |
MDL Number | MFCD00034667 |
InChI Key | FYGRPGOHQCPZCV-UHFFFAOYSA-N |
PubChem CID | 344887 |
IUPAC Name | ethyl 2-cyano-2-methylpropanoate |
SMILES | CCOC(=O)C(C)(C)C#N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15434087
|
Thermo Scientific Alfa Aesar
H31432.09 |
10 g |
42.21 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15444087
|
Thermo Scientific Alfa Aesar
H31432.18 |
50 g |
140.70 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is employed as a intermediates for pharmaceutical and organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is employed as a intermediates for pharmaceutical and organic synthesis.
Solubility
Soluble in ether. Not miscible or difficult to mix with water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Chemical Identifiers
1572-98-1 | |
141.17 | |
FYGRPGOHQCPZCV-UHFFFAOYSA-N | |
ethyl 2-cyano-2-methylpropanoate |
C7H11NO2 | |
MFCD00034667 | |
344887 | |
CCOC(=O)C(C)(C)C#N |
Specifications
1572-98-1 | |
185°C | |
MFCD00034667 | |
Soluble in ether. Not miscible or difficult to mix with water. | |
CCOC(=O)C(C)(C)C#N | |
141.17 | |
141.17 | |
Ethyl 2-cyano-2-methylpropionate |
0.97 | |
C7H11NO2 | |
2-cyano-2-methylpropionic acid ethyl ester, ethyl 2-cyano-2-methylpropionate, ethyl 2-cyano-2,2-dimethylacetate, 2-cyano-2-methylpropanoic acid ethyl ester, ethyl 2-cyano-2-methyl-propanoate, propanoic acid, 2-cyano-2-methyl-, ethyl ester, pubchem23352, acmc-209dfv, ksc529g3h, ethyl2-cyano-2-methylpropionate | |
FYGRPGOHQCPZCV-UHFFFAOYSA-N | |
ethyl 2-cyano-2-methylpropanoate | |
344887 | |
97% |
Safety and Handling
GHS H Statement
H301
Toxic if swallowed.
P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P501c
H302-H318
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.