missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-ethoxypropionate, 99+%

Product Code. p-7027347
Click to view available options
Quantity:
250 mL
1000 mL
Unit Size:
1000mL
250mL
This item is not returnable. View return policy

Product Code. 11416946

Brand: Thermo Scientific Alfa Aesar B24024.AK

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 763-69-9 | C7H14O3 | 146.186 g/mol

Ethyl 3-ethoxypropionate is used in the synthesis of phenols and selective inhibitors of cyclin-dependant kinase 4/6 for novel cancer therapies. It is also used as a solvent to prepare polymers. Further, it is used in paints and coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 3-ethoxypropionate is used in the synthesis of phenols and selective inhibitors of cyclin-dependant kinase 4/6 for novel cancer therapies. It is also used as a solvent to prepare polymers. Further, it is used in paints and coatings.

Solubility
Miscible with most organic solvents. Slightly miscible with cold water.

Notes
Incompatible with strong oxidizing agents, metals and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 763-69-9
Molecular Formula C7H14O3
Molecular Weight (g/mol) 146.186
MDL Number MFCD00051356
InChI Key BHXIWUJLHYHGSJ-UHFFFAOYSA-N
Synonym ethyl 3-ethoxypropionate, 3-ethoxypropionic acid ethyl ester, propanoic acid, 3-ethoxy-, ethyl ester, eep solvent, ethoxypropionic acid, ethyl ester, ethyl beta-ethoxypropionate, unii-ec38rsj79j, propionic acid, 3-ethoxy-, ethyl ester, ethyl-3-ethoxypropionate, ethylester kyseliny 3-ethoxypropionove
PubChem CID 12989
IUPAC Name ethyl 3-ethoxypropanoate
SMILES CCOCCC(=O)OCC

Specifications

Melting Point -100°C
Boiling Point 169°C to 171°C
Flash Point 59°C (142°F)
Odor Ester-like
Linear Formula CH3CH2OCH2CH2CO2CH2CH3
Quantity 250 mL
UN Number UN3272
Formula Weight 146.19
Percent Purity ≥99%
Chemical Name or Material Ethyl 3-ethoxypropionate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.